Spectra Por S/P 3 3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 33 mm dia; pack of 50.
Representative image only.
Spectra Por S/P 3 3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 33 mm dia; pack of 50.
Spectra Por | Mfr: 132488| Item: 0289066
Available in a variety of dimensions to meet your application needs
₹60,200.00INR / Pkg of 50
₹60,200.00INR / Pkg of 50
Buy It Together
Current Item
Spectra Por S/P 3 3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 33 mm dia; pack of 50.
Spectra Por S/P 3 3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 33 mm dia; pack of 50.
0289066
₹60,200.00INR / Pkg of 50
+ =
Total Price 105000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 3500 Dalton
1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)3500
  • Disc Diameter (mm)33
  • Description3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 33 mm dia; pack of 50.