Spectra-Por Micro Float-A-Lyzer Dialysis Device
Disposable ready-to-use dialysis device for micro-analysis
  • 98% sample recovery
  • Leak-proof luer-lok™ cap for sample protection
  • Available in 2 volume sizes
The Micro Float-A-Lyzer features ultrapure biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in seven precise MWCO's from 100 to 100,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included syringe. The sample device is self-floating due to an air bladder to maintain vertical orientation during dialysis; the sample device is also self standing for easy sample loading and retrieval. Individual units have interlocking housing for multi-sample dialysis. All sizes are supplied dry with glycerol in a package of 12.
37,300.00 - ₹43,500.00INR / Pkg of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Max Sample Volume (mL) Min Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por F235049 Micro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL 0290673 Mfr # F235049
COMPARE
500 0.2 100
₹37,300.00INR / Pkg of 12
Spectra Por F235051 Micro Float-A-Lyzer 0.5-1.0 kDalton, 100-200µL 0290674 Mfr # F235051
COMPARE
1000 0.2 500
₹37,300.00INR / Pkg of 12
Spectra Por F235053 Micro Float-A-Lyzer 3.5-5.0 kDalton, 100-200µL 0290675 Mfr # F235053
COMPARE
5000 0.2 3500
₹37,300.00INR / Pkg of 12
Spectra Por F235055 Micro Float-A-Lyzer 8.0-10.0 kDalton, 100-200µL 0290676 Mfr # F235055
COMPARE
10000 0.2 8000
₹37,300.00INR / Pkg of 12
Spectra Por F235058 Micro Float-A-Lyzer 50.0 kDalton, 100-200µL 0290678 Mfr # F235058
COMPARE
50000 0.2 50000
₹43,500.00INR / Pkg of 12
Spectra Por F235059 Micro Float-A-Lyzer 100.0 kDalton, 100-200µL 0290679 Mfr # F235059
COMPARE
100000 0.2 100000
₹37,300.00INR / Pkg of 12
Spectra Por F235061 Micro Float-A-Lyzer 0.1-0.5 kDalton, 400-500µL 0290683 Mfr # F235061
COMPARE
500 0.5 100
₹37,300.00INR / Pkg of 12
Spectra Por F235063 Micro Float-A-Lyzer 0.5-1.0 kDalton, 400-500µL 0290684 Mfr # F235063
COMPARE
1000 0.5 500
₹37,300.00INR / Pkg of 12
Spectra Por F235065 Micro Float-A-Lyzer 3.5-5.0 kDalton, 400-500µL 0290685 Mfr # F235065
COMPARE
5000 0.5 3500
₹37,300.00INR / Pkg of 12
Spectra Por F235067 Micro Float-A-Lyzer 8.0-10.0 kDalton, 400-500µL 0290686 Mfr # F235067
COMPARE
10000 0.5 8000
₹37,300.00INR / Pkg of 12
Spectra Por F235070 Micro Float-A-Lyzer 50.0 kDalton, 400-500µL 0290688 Mfr # F235070
COMPARE
50000 0.5 50000
₹37,300.00INR / Pkg of 12
Spectra Por F235071 Micro Float-A-Lyzer 100.0 kDalton, 400-500µL 0290689 Mfr # F235071
COMPARE
100000 0.5 100000
₹43,500.00INR / Pkg of 12
Showing 1- 12 of 12