Spectra Por F235071 Micro Float-A-Lyzer 100.0 kDalton, 400-500µL
Representative image only.
Spectra Por F235071 Micro Float-A-Lyzer 100.0 kDalton, 400-500µL
Spectra Por | Mfr: F235071| Item: 0290689
Disposable ready-to-use dialysis device for micro-analysis
₹43,500.00INR / Pkg of 12
₹43,500.00INR / Pkg of 12
Buy It Together
Current Item
Spectra Por F235071 Micro Float-A-Lyzer 100.0 kDalton, 400-500µL
Spectra Por F235071 Micro Float-A-Lyzer 100.0 kDalton, 400-500µL
0290689
₹43,500.00INR / Pkg of 12
+ + =
Total Price 180200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 98% sample recovery
  • Leak-proof luer-lok™ cap for sample protection
  • Available in 2 volume sizes
  • 1 year warranty
More About this Item
The Micro Float-A-Lyzer features ultrapure biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in seven precise MWCO's from 100 to 100,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included syringe. The sample device is self-floating due to an air bladder to maintain vertical orientation during dialysis; the sample device is also self standing for easy sample loading and retrieval. Individual units have interlocking housing for multi-sample dialysis. All sizes are supplied dry with glycerol in a package of 12.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)100000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)100000
  • Max Sample Volume (mL)0.5
  • Min Sample Volume (mL)0.4
  • ColorBlue
  • DescriptionMicro Float-A-Lyzer 100.0 kDalton, 400-500µL