Spectra Por F235049 Micro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL

Representative image only.

Spectra Por F235049 Micro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL

Spectra Por | Mfr: F235049| Item: 0290673

Disposable ready-to-use dialysis device for micro-analysis

₹37,300.00INR / Pkg of 12

₹37,300.00INR / Pkg of 12

Buy It Together

Current Item

Spectra Por F235049 Micro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL
Spectra Por F235049 Micro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL
0290673

₹37,300.00INR / Pkg of 12
+ + =
Total Price 174000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 98% sample recovery
  • Leak-proof luer-lok™ cap for sample protection
  • Available in 2 volume sizes
  • 1 year warranty

More About this Item

The Micro Float-A-Lyzer features ultrapure biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in seven precise MWCO's from 100 to 100,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included syringe. The sample device is self-floating due to an air bladder to maintain vertical orientation during dialysis; the sample device is also self standing for easy sample loading and retrieval. Individual units have interlocking housing for multi-sample dialysis. All sizes are supplied dry with glycerol in a package of 12.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)500
  • Min Molecular Weight Cut Off (Mwco) (Dalton)100
  • Max Sample Volume (mL)0.2
  • Min Sample Volume (mL)0.1
  • ColorGreen
  • DescriptionMicro Float-A-Lyzer 0.1-0.5 kDalton, 100-200µL
  • Warranty1 year
Reviews
Q&A