Safetec 15201 Multipurpose Spill, Chemical Clean-Up Kit
Representative image only.
Safetec Mfr # 15201 – Item # WW-86105-02
Portable for use by mobile response teams
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMultipurpose Spill, Chemical Clean-Up Kit
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Kit includes, barricade tape, one pair of heavy-duty gloves, pick-up scoop with scraper, respirator, safety goggles, ZAMMIE SEWERSTOPPERª, disposal bags (3), PETRO LOCKª, GREEN Zª, ACID LOCKª, ALKYª, hazardous material accident reporting kit.
Compliance: DOT 171.3 Hazardous Discharge, EPA 263.30 Immediate Action, and 263.31 Discharge Clean-up.