Mobile Spill Kits
Arrest spills anytime-anywhere
  • Wheeled bin for easy portability to every corner of the factory
  • Suitable for medium-scale spills
  • Effectively prevent and control possible liquid splashes, spills, or
  • pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
39,000.00 - ₹42,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chemical Treated Length (ft) Width (in)
Availability Pricing
SYSBEL SKIT002G Mobile Spill Kit, Universal; 45 gal.(170 L) 1598593 Mfr # SKIT002G
COMPARE
₹42,300.00INR / Each
SYSBEL SKIT002Y Mobile Spill Kit, Chemicals; 45 gal.(170 L) 1598594 Mfr # SKIT002Y
COMPARE
₹42,300.00INR / Each
SYSBEL SKIT002W Mobile Spill Kit, Oil only; 45 gal.(170 L) 1598595 Mfr # SKIT002W
COMPARE
₹39,000.00INR / Each