Clift Industries 3901-001 Mercury Eater Safety Spill Kit; 1 Gal.; 1 Pack/ Case
Representative image only.
Clift Industries 3901-001 Mercury Eater Safety Spill Kit; 1 Gal.; 1 Pack/ Case
Clift Industries | Mfr: 3901-001| Item: 8629506
Provides a quick, effective and safe method of handling all sizes of mercury spills
₹25,600.00INR / Each
₹25,600.00INR / Each
Product Description
Includes
1,800 grams of Mercury Eater, Mercury Eater final wipes, goggles, 1 pair of 18 mil latex gloves, whisk broom w/ dust pan, disposal bags, and instructions
Key Features
  • 2 year warranty
More About this Item
This Multi use self-contained safety spill kit has everything needed to contain, remove and decontaminate at all stages of mercury spill clean-up. Mercury Eater transforms mercury into a zinc amalgam which can be disposed of in a land-fill. Material removes mercury from work surfaces including bench tops, floors and hard-to-reach areas such as cracks and crevices. It absorbs visible mercury while suppressing the hazardous vapors. Kit comes with easy to step by step directions, in a sturdy, recognizable, re-usable container. Container has an easy entry screw-off top.
Two year warranty
Specifications
  • DescriptionMercury Eater Safety Spill Kit; 1 Gal.; 1 Pack/ Case