Drum Overpack Spill Kits
Arrest spills anytime-anywhere
  • Everything you need to clean up and dispose of a large-scale spill
  • Spill barrel prevents secondary leakage hazard
  • Contents are packed in a drum for easy portability and
  • disposal of materials
  • Effectively prevent and control possible liquid splashes, spills, or
  • pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
29,500.00 - ₹91,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Chemical Treated Length (yd) Width (in)
Availability Pricing
SYSBEL SYK950 Drum Overpack Spill Kit, Universal; 80 gal.(302 L) 1598584 Mfr # SYK950
COMPARE
₹90,400.00INR / Each
SYSBEL SYK951 Drum Overpack Spill Kit, Chemicals; 80 gal.(302 L) 1598585 Mfr # SYK951
COMPARE
₹91,800.00INR / Each
SYSBEL SYK952 Drum Overpack Spill Kit, Oil only; 80 gal.(302 L) 1598586 Mfr # SYK952
COMPARE
₹83,500.00INR / Each
SYSBEL SYK200 Drum Overpack Spill Kit, Universal; 21.5 gal.(81 L) 1598587 Mfr # SYK200
COMPARE
₹31,200.00INR / Each
SYSBEL SYK201 Drum Overpack Spill Kit, Chemicals; 21.5 gal.(81 L) 1598588 Mfr # SYK201
COMPARE
₹32,200.00INR / Each
SYSBEL SYK202 Drum Overpack Spill Kit, Oil only; 21.5 gal.(81 L) 1598589 Mfr # SYK202
COMPARE
₹29,500.00INR / Each