SYSBEL SYK952 Drum Overpack Spill Kit, Oil only; 80 gal.(302 L)
Representative image only.
SYSBEL SYK952 Drum Overpack Spill Kit, Oil only; 80 gal.(302 L)
SYSBEL | Mfr: SYK952| Item: 1598586
Arrest spills anytime-anywhere
₹83,500.00INR / Each
₹83,500.00INR / Each
Product Description
Key Features
  • Everything you need to clean up and dispose of a large-scale spill
  • Spill barrel prevents secondary leakage hazard
  • Contents are packed in a drum for easy portability and
  • disposal of materials
  • Effectively prevent and control possible liquid splashes, spills, or
  • pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
  • 15 month warranty
More About this Item
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
15 Month Warranty
Specifications
  • Absorption80 gal.(302 L)
  • ColorWhite
  • DescriptionDrum Overpack Spill Kit; Oil only
  • Warranty15 month