Portable Spill Kits
Arrest spills anytime-anywhere
  • Convenient kits in high-visibility yellow carrying bag
  • Suitable for small spills or leaks
  • Effectively prevent and control possible liquid splashes, spills, or pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
8,500.00 - ₹9,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chemical Treated Length (in) Width (in)
Availability Pricing
SYSBEL SKIT001G Portable Spill Kit, Universal; 9 to 13 gal.(35 to 50 L) WW-15985-96 Mfr # SKIT001G
COMPARE
₹9,200.00INR / Each
SYSBEL SKIT001Y Portable Spill Kit, Chemicals; 9 to 13 gal.(35 to 50 L) WW-15985-97 Mfr # SKIT001Y
COMPARE
₹9,200.00INR / Each
SYSBEL SKIT001W Portable Spill Kit, Oil only; 9 to 13 gal.(35 to 50 L) WW-15985-98 Mfr # SKIT001W
COMPARE
₹8,500.00INR / Each