SYSBEL SKIT001W Portable Spill Kit, Oil only; 9 to 13 gal.(35 to 50 L)
Representative image only.
SYSBEL SKIT001W Portable Spill Kit, Oil only; 9 to 13 gal.(35 to 50 L)
SYSBEL | Mfr: SKIT001W| Item: 1598598
Arrest spills anytime-anywhere
₹7,600.00INR / Each
₹7,600.00INR / Each
Product Description
Key Features
  • Convenient kits in high-visibility yellow carrying bag
  • Suitable for small spills or leaks
  • Effectively prevent and control possible liquid splashes, spills, or pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
  • 15 month warranty
More About this Item
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
15 Month Warranty
Specifications
  • Absorption9 to 13 gal.(35 to 50 L)
  • ColorWhite
  • DescriptionPortable Spill Kit; Oil only
  • Warranty15 month