Absorbents

Pads are for small spills; use to cover the spill area until the spills
are completely absorbed
Socks absorb and contain spill range by placing in an
overlapping circle
Pillows are ideal for dealing with waste tanks, coolant tanks,
and storage containers
  • Spill control products are essential to any well-organized lab. When
  • spills happen in the workplace, the type of spill needs to be quickly
  • identified and the correct products used for spill containment
  • and cleanup. Universal (gray): oils, coolant, solvents, and water.
  • Chemicals (yellow): acids, corrosives, and unknown hazardous
  • chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
  • not water.
  • Melt-blow polypropylene (MBPP) absorbent materials feature high
  • absorption rate, rapid absorption, good cleaning effect; don’t react
  • with polar, toxic, hazardous chemicals; and are safe to use, causing
  • no harm to personal health.
9,800.00 - ₹16,100.00INR / Each

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Absorption Chemical Treated Description
Availability Pricing
SYSBEL UP0001G Absorbent Pads, Universal; 16 to 26 gal.(60 to 100 L) WW-15985-99 Mfr # UP0001G
COMPARE
16 to 26 gal.(60 to 100 L) Absorbent Pads; Universal
₹11,100.00INR / Each
SYSBEL CP0001Y Absorbent Pads, Chemicals; 16 to 26 gal.(60 to 100 L) WW-15986-00 Mfr # CP0001Y
COMPARE
16 to 26 gal.(60 to 100 L) Absorbent Pads; Chemicals
₹11,800.00INR / Each
SYSBEL OP0001W Absorbent Pads, Oil only; 16 to 26 gal.(60 to 100 L) WW-15986-01 Mfr # OP0001W
COMPARE
16 to 26 gal.(60 to 100 L) Absorbent Pads; Oil only
₹9,800.00INR / Each
SYSBEL US0001G Absorbent Socks, Universal; 18 to 26 gal.(70 to 100 L) WW-15986-02 Mfr # US0001G
COMPARE
18 to 26 gal.(70 to 100 L) Absorbent Socks; Universal
₹14,100.00INR / Each
SYSBEL CS0001Y Absorbent Socks, Chemicals; 18 to 26 gal.(70 to 100 L) WW-15986-03 Mfr # CS0001Y
COMPARE
18 to 26 gal.(70 to 100 L) Absorbent Socks; Chemicals
₹16,100.00INR / Each
SYSBEL OS0001W Absorbent Socks, Oil only; 18 to 26 gal.(70 to 100 L) WW-15986-04 Mfr # OS0001W
COMPARE
18 to 26 gal.(70 to 100 L) Absorbent Socks; Oil only
₹13,800.00INR / Each
SYSBEL SUP001 Absorbent Pillow, Universal; 21 to 29 gal.(80 to 110 L) WW-15986-05 Mfr # SUP001
COMPARE
21 to 29 gal.(80 to 110 L) Absorbent Pillow; Universal
₹13,400.00INR / Each
SYSBEL SCP001 Absorbent Pillow, Chemicals; 21 to 29 gal.(80 to 110 L) WW-15986-06 Mfr # SCP001
COMPARE
21 to 29 gal.(80 to 110 L) Absorbent Pillow; Chemicals
₹14,100.00INR / Each
SYSBEL SOP001 Absorbent Pillow, Oil only; 21 to 29 gal.(80 to 110 L) WW-15986-07 Mfr # SOP001
COMPARE
21 to 29 gal.(80 to 110 L) Absorbent Pillow; Oil only
₹11,500.00INR / Each
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat