SYSBEL CP0001Y Absorbent Pads, Chemicals; 16 to 26 gal.(60 to 100 L)
Representative image only.
SYSBEL CP0001Y Absorbent Pads, Chemicals; 16 to 26 gal.(60 to 100 L)
SYSBEL | Mfr: CP0001Y| Item: 1598600
Pads are for small spills; use to cover the spill area until the spills
are completely absorbed
Socks absorb and contain spill range by placing in an
overlapping circle
Pillows are ideal for dealing with waste tanks, coolant tanks,
and storage containers
₹10,400.00INR / Each
₹10,400.00INR / Each
Product Description
Key Features
  • Spill control products are essential to any well-organized lab. When
  • spills happen in the workplace, the type of spill needs to be quickly
  • identified and the correct products used for spill containment
  • and cleanup. Universal (gray): oils, coolant, solvents, and water.
  • Chemicals (yellow): acids, corrosives, and unknown hazardous
  • chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
  • not water.
  • Melt-blow polypropylene (MBPP) absorbent materials feature high
  • absorption rate, rapid absorption, good cleaning effect; don’t react
  • with polar, toxic, hazardous chemicals; and are safe to use, causing
  • no harm to personal health.
  • 15 month warranty
15 Month Warranty
Specifications
  • Absorption16 to 26 gal.(60 to 100 L)
  • ColorYellow
  • DescriptionAbsorbent Pads; Chemicals
  • Warranty15 month