Eagle Modular Spill Containment Platforms
Connect multiple units for custom layout options
  • Grating removes easily for simple clean-up
  • Meet EPS 40 CFR 264.175
  • SPCC Cmpliant
The modular spill containment platforms are constructed of high-density polyethylene (HDPE) for excellent durability and chemical resistance. Sump capacities between 15 gallons, measures only 6.5" high and has black HDPE flat top grating which removes easily for cleaning. Secure platforms together with built-in connections. Order optional ramp to attach to platform system for easy loading and unloading.
34,800.00 - ₹81,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Description
Availability Pricing
Eagle 1633 Modular Spill Containment Platform without Drain, 1 Drum, 15 Gal Sump Capacity 8611009 Mfr # 1633
COMPARE
12 Modular Spill Control Platform; 1-drum, 12-gallon capacity
₹34,800.00INR / Each
Eagle 1632 Modular Spill Containment Platform without Drain, 2 Drums, 30 Gal Sump Capacity 8611011 Mfr # 1632
COMPARE
30 Modular Spill Control Platform; 2-drum, 30-gallon capacity
₹63,800.00INR / Each
Eagle 1645 Modular Spill Containment Platform with Drain, 4 Drums, 66 Gal Sump Capacity 8611014 Mfr # 1645
COMPARE
66 Spill Control Platform; 4-Drum, 66-gallon Capacity
₹81,900.00INR / Each