Scienceware F36250 Mercury Collector Jar with Integrated Sponge and Perforated Plate
Representative image only.
Scienceware F36250 Mercury Collector Jar with Integrated Sponge and Perforated Plate
Scienceware | Mfr: F36250-0000| Item: 0670600
Makes small mercury spill clean-up quick and simple
₹10,000.00INR / Each
₹10,000.00INR / Each
Product Description
More About this Item
Foam pad on lid liner absorbs spilled mercury. Simply screw on lid to collect mercury beneath perforated plate.
Specifications
  • OD (in)4 1/8
  • Height (in)2
  • DescriptionMercury Collector Jar with Integrated Sponge and Perforated Plate