QLA Standard Sampling Probe Top for Distek Baths
Representative image only.
QLA Standard Sampling Probe Top for Distek Baths
| Mfr: CANTOP-DK| Item: 4130283
Achieve consistent sampling with QLA Distek-compatible probes
₹27,000.00INR / Each
₹27,000.00INR / Each
Buy It Together
Current Item
QLA Standard Sampling Probe Top for Distek Baths
QLA Standard Sampling Probe Top for Distek Baths
4130283
₹27,000.00INR / Each
+ =
Total Price 43600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Sampling probe top
  • 1/16" stopper
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • DescriptionStandard Sampling Probe Top for Distek Baths