QLA Distek/Hanson Cannula Stopper for VanKel EaseAlign Cover; 1/16" (1.6 mm) OD
Representative image only.
QLA Distek/Hanson Cannula Stopper for VanKel EaseAlign Cover; 1/16" (1.6 mm) OD
| Mfr: CANSTP-065-31| Item: 4130267
QLA accessories help provide more precise and consistent dissolution results
₹18,100.00INR / Each
₹18,100.00INR / Each
Product Description
Key Features
  • Distek/Hanson stopper for VanKel EaseAlign cover
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithVanKel EaseAlign Cover
  • DescriptionDistek/Hanson Cannula Stopper for VanKel EaseAlign Cover; 1/16" (1.6 mm) OD