QLA Hanson Cannula Stopper for Distek Cover; 1/8" (3.2 mm) OD
Representative image only.
QLA Hanson Cannula Stopper for Distek Cover; 1/8" (3.2 mm) OD
| Mfr: CANSTP-125-53| Item: 4130273
QLA accessories help provide more precise and consistent dissolution results
₹17,000.00INR / Each
₹17,000.00INR / Each
Product Description
Key Features
  • Hanson stopper for Distek cover
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithDistek cover
  • DescriptionHanson Cannula Stopper for Distek Cover; 1/8" (3.2 mm) OD