QLA Adjustable Cannula Stopper for Distek; 1/8" (3.2 mm) OD
Representative image only.
QLA Adjustable Cannula Stopper for Distek; 1/8" (3.2 mm) OD
| Mfr: CA8STP-DK| Item: 4130208
QLA accessories help provide more precise and consistent dissolution results
₹14,100.00INR / Each
₹14,100.00INR / Each
Product Description
Key Features
  • For Distek
  • 1/8" (3.2 mm) OD
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithDistek apparatus
  • DescriptionAdjustable Cannula Stopper for Distek; 1/8" (3.2 mm) OD