QLA VanKel Cannula Stopper for Distek Cover
Representative image only.
QLA VanKel Cannula Stopper for Distek Cover
| Mfr: CANSTP-078-53| Item: 4130270
QLA accessories help provide more precise and consistent dissolution results
₹17,000.00INR / Each
₹17,000.00INR / Each
Product Description
Key Features
  • VanKel stopper for Distek cover
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithDistek cover
  • DescriptionVanKel Cannula Stopper for Distek Cover