QLA Adjustable Cannula Stopper for Distek, 0.082" OD; with Set Screw
Representative image only.
QLA Adjustable Cannula Stopper for Distek, 0.082" OD; with Set Screw
| Mfr: CANSTP-DK | Item: 4130277
QLA accessories help provide more precise and consistent dissolution results
₹19,000.00INR / Each
₹19,000.00INR / Each
Product Description
Key Features
  • For Distek
  • 0.082" OD
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithDistek apparatus
  • DescriptionAdjustable Cannula Stopper for Distek, 0.082" OD; with Set Screw