QLA Distek Cannula Stopper with PTFE Sleeve for VanKel EaseAlign Cover; 1/8" (3.2 mm) OD
Representative image only.
QLA Distek Cannula Stopper with PTFE Sleeve for VanKel EaseAlign Cover; 1/8" (3.2 mm) OD
| Mfr: CA9STP-DV| Item: 4130210
QLA accessories help provide more precise and consistent dissolution results
₹16,600.00INR / Each
₹16,600.00INR / Each
Product Description
Key Features
  • Distek stopper with PTFE sleeve for VanKel EaseAlign cover
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithVanKel EaseAlign Cover
  • DescriptionDistek Cannula Stopper with PTFE Sleeve for VanKel EaseAlign Cover; 1/8" (3.2 mm) OD