QLA Standard Sampling Probe Bottom for Distek Baths; 500 mL
Representative image only.
QLA Standard Sampling Probe Bottom for Distek Baths; 500 mL
| Mfr: CAN5BM-DK| Item: 4130243
Achieve consistent sampling with QLA Distek-compatible probes
₹26,100.00INR / Each
₹26,100.00INR / Each
Buy It Together
Current Item
QLA Standard Sampling Probe Bottom for Distek Baths; 500 mL
QLA Standard Sampling Probe Bottom for Distek Baths; 500 mL
4130243
₹26,100.00INR / Each
+ =
Total Price 42700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Sampling probe bottom
  • Use with 500 mL vessel
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • Compatible With500 mL vessel
  • DescriptionStandard Sampling Probe Bottom for Distek Baths; 500 mL