QLA Solid Housing Sampling Probe for Distek Baths; 500 mL
Representative image only.
QLA Solid Housing Sampling Probe for Distek Baths; 500 mL
| Mfr: CAN500-DSH| Item: 4130240
Achieve consistent sampling with QLA Distek-compatible probes
₹44,100.00INR / Each
₹44,100.00INR / Each
Buy It Together
Current Item
QLA Solid Housing Sampling Probe for Distek Baths; 500 mL
QLA Solid Housing Sampling Probe for Distek Baths; 500 mL
4130240
₹44,100.00INR / Each
+ =
Total Price 60700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Solid housing sampling probe
  • Use with 500 mL vessel
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • Compatible With500 mL vessel
  • DescriptionSolid Housing Sampling Probe for Distek Baths; 500 mL