QLA Return Probe for Distek Baths
Representative image only.
QLA Return Probe for Distek Baths
| Mfr: CANRET-DK| Item: 4130264
Achieve consistent sampling with QLA Distek-compatible probes
₹28,000.00INR / Each
₹28,000.00INR / Each
Buy It Together
Current Item
QLA Return Probe for Distek Baths
QLA Return Probe for Distek Baths
4130264
₹28,000.00INR / Each
+ =
Total Price 44600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Return probe
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • DescriptionReturn Probe for Distek Baths