QLA DV Series Sampling Probe for Distek Baths; 900 mL
Representative image only.
QLA DV Series Sampling Probe for Distek Baths; 900 mL
| Mfr: CAN900-DV| Item: 4130259
Achieve consistent sampling with QLA Distek-compatible probes
₹46,100.00INR / Each
₹46,100.00INR / Each
Buy It Together
Current Item
QLA DV Series Sampling Probe for Distek Baths; 900 mL
QLA DV Series Sampling Probe for Distek Baths; 900 mL
4130259
₹46,100.00INR / Each
+ =
Total Price 62700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • DV-series sampling probe
  • Use with 900 mL vessel
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • Compatible With900 mL vessel
  • DescriptionDV Series Sampling Probe for Distek Baths; 900 mL