QLA Vessel Covers
Reduce loss of media due to evaporation
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
7,500.00 - ₹86,100.00INR / Each
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
QLA Hinged Cover for Stationary Basket, Clear, with Extra Hole; for Distek Baths 4130805 Mfr # COVDHG-SB
COMPARE
Clear Distek baths Hinged Cover for Stationary Basket, Clear, with Extra Hole; for Distek Baths
₹45,600.00INR / Each
QLA Hinged Cover for Easy Basket Access, Amber, with Large Center Hole; for Distek Baths 4130806 Mfr # COVERAD-BK
COMPARE
Amber Distek baths Hinged Cover for Easy Basket Access, Amber, with Large Center Hole; for Distek Baths
₹75,100.00INR / Each
QLA Low Evaporation Rotary Cover for Distek Baths 4130807 Mfr # COVERD-00
COMPARE
White Distek baths Low Evaporation Rotary Cover for Distek Baths
₹36,500.00INR / Each
QLA Hinged Cover, Clear; for Distek Baths 4130808 Mfr # COVERD-01
COMPARE
Clear Distek baths Hinged Cover, Clear; for Distek Baths
₹33,100.00INR / Each
QLA Hinged Cover, Clear, with Paddle Slot; for ACCUBKT-VK & VK Auto-Sampler 4130809 Mfr # COVERD-01VKBKT
COMPARE
Clear Agilent ACCUBKT-VK & VK auto sampler Hinged Cover, Clear, with Paddle Slot; for ACCUBKT-VK & VK Auto-Sampler
₹42,000.00INR / Each
QLA Hinged Cover, Amber; for Distek Baths 4130810 Mfr # COVERD-02
COMPARE
Amber Distek baths Hinged Cover, Amber; for Distek Baths
₹41,100.00INR / Each
QLA Hinged Cover, Clear, 0.125" Sampling Hole; For Distek Baths 4130811 Mfr # COVERD-033
COMPARE
Clear Distek baths Hinged Cover, Clear, 0.125" Sampling Hole; For Distek Baths
₹30,200.00INR / Each
QLA Hinged Cover,Black Acrylic, with Small Center Hole; for Distek Baths 4130812 Mfr # COVERD-04
COMPARE
Black Distek baths Hinged Cover,Black Acrylic, with Small Center Hole; for Distek Baths
₹65,000.00INR / Each
QLA Hinged Cover for Easy Basket Access, Clear, with Large Center Hole; for Distek Baths 4130813 Mfr # COVERD-BK
COMPARE
Clear Distek baths Hinged Cover for Easy Basket Access, Clear, with Large Center Hole; for Distek Baths
₹52,100.00INR / Each
QLA Hinged Cover, Clear, Low Evaporation; for Erweka Baths 4130814 Mfr # COVERD-EW
COMPARE
Clear Erweka baths Hinged Cover, Clear, Low Evaporation; for Erweka Baths
₹34,200.00INR / Each
QLA Hinged Cover, Clear, Low Evaporation; for Distek Baths 4130815 Mfr # COVERD-HG
COMPARE
Clear Distek baths Hinged Cover, Clear, Low Evaporation; for Distek Baths
₹29,100.00INR / Each
QLA Rotary Cover, Low Evaporation; for Hanson SR8-Plus Baths 4130816 Mfr # COVERH-00
COMPARE
White Hanson SR8-Plus baths Rotary Cover, Low Evaporation; for Hanson SR8-Plus Baths
₹41,600.00INR / Each
QLA Cover with Paddle Slot, Clear; for Hanson SR8-Plus Baths 4130817 Mfr # COVERH-01
COMPARE
Clear Hanson SR8-Plus baths Cover with Paddle Slot, Clear; for Hanson SR8-Plus Baths
₹14,000.00INR / Each
QLA Cover with Paddle Slot, Gray; for Hanson SR8-Plus Baths 4130818 Mfr # COVERH-02
COMPARE
Gray Hanson SR8-Plus baths Cover with Paddle Slot, Gray; for Hanson SR8-Plus Baths
₹12,500.00INR / Each
QLA Cover with Paddle Slot, Amber; for Hanson SR8-Plus Baths 4130819 Mfr # COVERH-03
COMPARE
Amber Hanson SR8-Plus baths Cover with Paddle Slot, Amber; for Hanson SR8-Plus Baths
₹7,500.00INR / Each
QLA Solid Cover, Clear; for Hanson SR8-Plus Baths 4130820 Mfr # COVERH-04
COMPARE
Clear Hanson SR8-Plus baths Solid Cover, Clear; for Hanson SR8-Plus Baths
₹18,100.00INR / Each
QLA Hinged Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths 4130821 Mfr # COVERH-HG
COMPARE
Clear Hanson SR8-Plus baths Hinged Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths
₹33,500.00INR / Each
QLA Conical Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths 4130822 Mfr # COVERH-LL
COMPARE
Clear Hanson SR8-Plus baths Conical Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths
₹74,600.00INR / Each
QLA Rotary Cover,Low Evaporation; for VanKel EaseAlign Bracket 4130823 Mfr # COVERV-00
COMPARE
White VanKel EaseAlign bracket Rotary Cover,Low Evaporation; for VanKel EaseAlign Bracket
₹44,500.00INR / Each
QLA Cover with Paddle Slot, Clear; for VanKel EaseAlign Bracket 4130824 Mfr # COVERV-01
COMPARE
Clear VanKel EaseAlign bracket Cover with Paddle Slot, Clear; for VanKel EaseAlign Bracket
₹26,000.00INR / Each
Showing 1 - 20 of 34