QLA Hinged Cover for Stationary Basket, Clear, with Extra Hole; for Distek Baths
Representative image only.
QLA Hinged Cover for Stationary Basket, Clear, with Extra Hole; for Distek Baths
| Mfr: COVDHG-SB| Item: 4130805
Reduce loss of media due to evaporation
₹45,600.00INR / Each
₹45,600.00INR / Each
Product Description
Key Features
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
  • For Distek baths
  • Clear with extra hole
  • Hinged
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
.
Specifications
  • ColorClear
  • Compatible WithDistek baths
  • DescriptionHinged Cover for Stationary Basket, Clear, with Extra Hole; for Distek Baths