QLA Hinged Cover, Clear, Low Evaporation; for Erweka Baths
Representative image only.
QLA Hinged Cover, Clear, Low Evaporation; for Erweka Baths
| Mfr: COVERD-EW| Item: 4130814
Reduce loss of media due to evaporation
₹34,200.00INR / Each
₹34,200.00INR / Each
Product Description
Key Features
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
  • For Erweka baths
  • Hinged
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
.
Specifications
  • ColorClear
  • Compatible WithErweka baths
  • DescriptionHinged Cover, Clear, Low Evaporation; for Erweka Baths