QLA Hinged Cover, Clear, with Paddle Slot; for ACCUBKT-VK & VK Auto-Sampler
Representative image only.
QLA Hinged Cover, Clear, with Paddle Slot; for ACCUBKT-VK & VK Auto-Sampler
| Mfr: COVERD-01VKBKT| Item: 4130809
Reduce loss of media due to evaporation
₹42,000.00INR / Each
₹42,000.00INR / Each
Product Description
Key Features
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
  • For ACCUBKT-VK & VK auto-sampler
  • Clear with paddle slot
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
.
.
Specifications
  • ColorClear
  • Compatible WithAgilent ACCUBKT-VK & VK auto sampler
  • DescriptionHinged Cover, Clear, with Paddle Slot; for ACCUBKT-VK & VK Auto-Sampler