QLA Hinged Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths
Representative image only.
QLA Hinged Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths
| Mfr: COVERH-HG| Item: 4130821
Reduce loss of media due to evaporation
₹33,500.00INR / Each
₹33,500.00INR / Each
Product Description
Key Features
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
  • For Hanson SR8-Plus baths
  • Hinged
  • Low evaporation
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
.
Specifications
  • ColorClear
  • Compatible WithHanson SR8-Plus baths
  • DescriptionHinged Cover, Clear, Low Evaporation; for Hanson SR8-Plus Baths