QLA Rotary Cover, Low Evaporation; for Hanson SR8-Plus Baths
Representative image only.
QLA Rotary Cover, Low Evaporation; for Hanson SR8-Plus Baths
| Mfr: COVERH-00| Item: 4130816
Reduce loss of media due to evaporation
₹41,600.00INR / Each
₹41,600.00INR / Each
Product Description
Key Features
  • Protect samples from contamination or spillage Specific styles and colors assist almost every process
  • For Hanson SR8-Plus baths
  • Rotary design
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
.
Specifications
  • ColorWhite
  • Compatible WithHanson SR8-Plus baths
  • DescriptionRotary Cover, Low Evaporation; for Hanson SR8-Plus Baths