Kimble Disposable Culture Tubes
Ideal for general bacteriological and tissue culture applications
  • Tubes are offered with or without white markings
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.
52,300.00 - ₹102,000.00INR / Case of 1000
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Label Type Material
Availability Pricing
DWK Life Sciences (Kimble) 73750-13100 Disposable Culture Tubes, 13 x 100, with markings 1000/cs WW-34580-00 Mfr # 73750-13100
COMPARE
8 White Write-On Glass
₹78,200.00INR / Case of 1000
DWK Life Sciences (Kimble) 73750-16100 Disposable Culture Tubes, 16 x 100, with markings 1000/cs WW-34580-02 Mfr # 73750-16100
COMPARE
12 White Write-On Glass
₹76,000.00INR / Case of 1000
DWK Life Sciences (Kimble) 73750-16125 Disposable Culture Tubes, 16 x 125, with markings 1000/cs WW-34580-04 Mfr # 73750-16125
COMPARE
16 White Write-On Glass
₹87,300.00INR / Case of 1000
DWK Life Sciences (Kimble) 73750-16150 Disposable Culture Tubes, 16 x 150, with markings 1000/cs WW-34580-06 Mfr # 73750-16150
COMPARE
20 White Write-On Glass
₹77,700.00INR / Case of 1000
DWK Life Sciences (Kimble) 73750-20125 Disposable Culture Tubes, 20 x 125, with markings 500/cs WW-34580-08 Mfr # 73750-20125
COMPARE
25 White Write-On Glass
₹52,300.00INR / Case of 500
DWK Life Sciences (Kimble) 73750-20150 Disposable Culture Tubes, 20 x 150, with markings 500/cs WW-34580-10 Mfr # 73750-20150
COMPARE
30 White Write-On Glass
₹59,000.00INR / Case of 500
DWK Life Sciences (Kimble) 73770-13100 Disposable Round Bottom Culture Tubes, 13 x 100, no markings, 1000/cs WW-34580-50 Mfr # 73770-13100
COMPARE
8 None Glass
₹69,700.00INR / Case of 1000
DWK Life Sciences (Kimble) 73770-16100 Disposable Round Bottom Culture Tubes, 16 x 100, no markings, 1000/cs WW-34580-52 Mfr # 73770-16100
COMPARE
12 None Glass
₹102,000.00INR / Case of 1000
DWK Life Sciences (Kimble) 73770-16125 Disposable Round Bottom Culture Tubes, 16 x 125, no markings, 1000/cs WW-34580-54 Mfr # 73770-16125
COMPARE
16 None Glass
₹84,900.00INR / Case of 1000
DWK Life Sciences (Kimble) 73770-16150 Disposable Round Bottom Culture Tubes, 16 x 150, no markings, 1000/cs WW-34580-56 Mfr # 73770-16150
COMPARE
20 None Glass
₹91,000.00INR / Case of 1000
DWK Life Sciences (Kimble) 73770-20125 Disposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs WW-34580-58 Mfr # 73770-20125
COMPARE
25 None Glass
₹63,000.00INR / Case of 500
DWK Life Sciences (Kimble) 73770-20150 Disposable Round Bottom Culture Tubes, 20 x 150, no markings, 500/cs WW-34580-60 Mfr # 73770-20150
COMPARE
30 None Glass
₹73,400.00INR / Case of 500
Showing 1- 12 of 12