DWK Life Sciences (Kimble) 73750-20125 Disposable Culture Tubes, 20 x 125, with markings 500/cs
Representative image only.
DWK Life Sciences (Kimble) 73750-20125 Disposable Culture Tubes, 20 x 125, with markings 500/cs
DWK Life Sciences (Kimble) | Mfr: 73750-20125| Item: 3458008
Ideal for general bacteriological and tissue culture applications
₹58,600.00INR / Case of 500
₹58,600.00INR / Case of 500
Buy It Together
Current Item
DWK Life Sciences (Kimble) 73750-20125 Disposable Culture Tubes, 20 x 125, with markings 500/cs
DWK Life Sciences (Kimble) 73750-20125 Disposable Culture Tubes, 20 x 125, with markings 500/cs
3458008
₹58,600.00INR / Case of 500
+ =
Total Price 138400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Tubes are offered with or without white markings
More About this Item
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.
Specifications
  • Capacity (mL)25
  • MaterialGlass
  • Cap / Stopper Size18-415
  • Label TypeWhite Write-On
  • AutoclavableYes
  • Outside Diameter (OD) (cm)2
  • Overall Height (cm)12.5
  • DescriptionDisposable Culture Tubes, 20 x 125, with markings 500/cs