DWK Life Sciences (Kimble) Phenolic Caps, Cemented Rubber Liner, 18-415 Threaded; 1000/Cs
Representative image only.
DWK Life Sciences (Kimble) Phenolic Caps, Cemented Rubber Liner, 18-415 Threaded; 1000/Cs
DWK Life Sciences (Kimble) | Mfr: 73800-18415| Item: 3458036
Snuggle fits Kimble Disposable Culture Tubes
₹79,800.00INR / Case of 1000
₹79,800.00INR / Case of 1000
Product Description
Key Features
  • Excellent for general lab use
  • Cemented-in rubber liner
  • Autoclavable
More About this Item
Keep your samples secure with these black phenolic screw on caps designed to fit your Kimble disposable culture tubes.
Specifications
  • MaterialPhenolic
  • ColorBlack
  • Compatible With34580-08, -10, 58, and -60
  • Connections18-415
  • DescriptionPhenolic Caps, Cemented Rubber Liner, 18-415 Threaded