DWK Life Sciences (Kimble) 73750-16150 Disposable Culture Tubes, 16 x 150, with markings 1000/cs
Representative image only.
DWK Life Sciences (Kimble) 73750-16150 Disposable Culture Tubes, 16 x 150, with markings 1000/cs
DWK Life Sciences (Kimble) | Mfr: 73750-16150| Item: 3458006
Ideal for general bacteriological and tissue culture applications
₹87,600.00INR / Case of 1000
₹87,600.00INR / Case of 1000
Product Description
Key Features
  • Tubes are offered with or without white markings
More About this Item
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.
Specifications
  • Capacity (mL)20
  • MaterialGlass
  • Cap / Stopper Size15-415
  • AutoclavableYes
  • Label TypeWhite Write-On
  • Outside Diameter (OD) (cm)1.6
  • Overall Height (cm)15
  • DescriptionDisposable Culture Tubes, 16 x 150, with markings 1000/cs