Environmental Express® TKN Standards and Reagents

Meet EPA method 351.1 and 351.2
  • Ready to use standards and reagents
  • Boiling chips keep heat steady
Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.
11,000.00 - ₹11,400.00INR / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
LabChem Copper sulfate pentahydrate, 500 g 3520536 Mfr # LC134051
COMPARE
Copper sulfate pentahydrate, 500g
Call For Price
LabChem Digestion Reagent (with Mercury), 1 L 3520537 Mfr # LC105502
COMPARE
Digestion Reagent (with Mercury), 1L
Call For Price
LabChem Digestion Reagent (without Mercury), 1 L 3520539 Mfr # LC105602
COMPARE
Digestion Reagent (without Mercury), 1L
Call For Price
COMPARE
Carbon Boiling Chips 8oz
₹11,000.00INR / Each
LabChem Optional EDTA 5% w/v, 500 mL 3520543 Mfr # LC139301
COMPARE
Optional EDTA 5% w/v, 500 mL
Call For Price
LabChem Sodium hypochlorite 2-3% w/v, 1 L 3520544 Mfr # LC246402
COMPARE
Sodium hypochlorite 2-3% w/v, 1L
Call For Price
LabChem Sodium nitroprusside 0.05% w/v, 500 mL 3520545 Mfr # LC247001
COMPARE
Sodium nitroprusside 0.05% w/v, 500 mL
₹11,400.00INR / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat