LabChem Copper sulfate pentahydrate, 500 g

Representative image only.

LabChem Copper sulfate pentahydrate, 500 g

| Mfr: LC134051| Item: 3520536

Meet EPA method 351.1 and 351.2

Call For Price

Call For Price
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Specifications
  • DescriptionCopper sulfate pentahydrate, 500g
Reviews
Q&A