LabChem Sodium hypochlorite 2-3% w/v, 1 L

Representative image only.

LabChem Sodium hypochlorite 2-3% w/v, 1 L

LabChem | Mfr: LC246402| Item: 3520544

Meet EPA method 351.1 and 351.2

Call For Price

Call For Price
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Specifications
  • DescriptionSodium hypochlorite 2-3% w/v, 1L
Reviews
Q&A