Carbon Boiling Chips 8 oz

Representative image only.

Carbon Boiling Chips 8 oz

| Item: 3520541

Meet EPA method 351.1 and 351.2

₹11,000.00INR / Each

₹11,000.00INR / Each
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Specifications
  • DescriptionCarbon Boiling Chips 8oz
Reviews
Q&A