LabChem Optional EDTA 5% w/v, 500 mL

Representative image only.

LabChem Optional EDTA 5% w/v, 500 mL

LabChem | Mfr: LC139301| Item: 3520543

Meet EPA method 351.1 and 351.2

Call For Price

Call For Price
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Specifications
  • DescriptionOptional EDTA 5% w/v, 500 mL
Reviews
Q&A