LabChem Digestion Reagent (with Mercury), 1 L

Representative image only.

LabChem Digestion Reagent (with Mercury), 1 L

LabChem | Mfr: LC105502| Item: 3520537

Meet EPA method 351.1 and 351.2

Call For Price

Call For Price
Product Description

Key Features

  • Ready to use standards and reagents
  • Boiling chips keep heat steady

More About this Item

Standards and reagents pre-made to meet your Total Kjeldahl Nitrogen (TKN) testing needs.

Warning: Contains mercury (Hg). Do not dispose of in household waste. Dispose of in accordance with applicable local, national, or international regulations.

Specifications
  • DescriptionDigestion Reagent (with Mercury), 1L
Reviews
Q&A