Environmental Express® Air Sampling Filters
Ideal for air particle monitoring and environmental testing
  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance
Quality filters to meet your industrial hygiene applications with excellent air permeability with minimal pressure drop.
4,000.00 - ₹153,000.00INR / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Description
Availability Pricing
COMPARE
25 Cellulose support pad Air Filter Cellulose Support Pads, White/25mm; 100/Pk
₹4,000.00INR / Pkg of 100
COMPARE
37 Cellulose support pad Air Sampling Filter Cellulose Support Pads, White Plain, 37mm; 100/Pk
₹4,600.00INR / Pkg of 100
COMPARE
25 Cellulose support pad Air Filter Cellulose Support Pads, Gridded, 25mm: 100/Pk
₹4,400.00INR / Pkg of 100
COMPARE
37 Cellulose support pad Air Filter Cellulose Support Pads, Gridded, 37mm; 100/Pk
₹5,400.00INR / Pkg of 100
COMPARE
203 Borosilicate Glass Microfiber Air Sampling Borosilicate GF Filters, 8" x 10" Sheets; 100/Pk
₹45,900.00INR / Pkg of 100
COMPARE
203 Quartz Microfiber Air Sampling Quartz Fiber Filters, 8" x 10" Sheets; 50/Pk
₹153,000.00INR / Pkg of 50
Advantec Air Sampling Binderless GF Filter, 25 mm, 1um; Ea 3520921 Mfr # FG120025
COMPARE
25 Borosilicate Glass Microfiber Air Sampling Binderless GF Filter, 25 mm, 1um; Ea
₹5,400.00INR / Pkg of 1
COMPARE
25 Mixed Cellulose Ester w/pad Air Sampling MCE Filters with Pads, 25 mm, 0.45 µm; 100/Pk
Call For Price
COMPARE
37 Mixed Cellulose Ester w/pad Air Sampling MCE Filters with Pads, 37 mm, 0.45 µm; 100/Pk
Call For Price
COMPARE
25 Mixed Cellulose Ester w/pad Air Sampling MCE Filters with Pads, 25 mm, 0.8 µm; 100/Pk
Call For Price
COMPARE
37 Mixed Cellulose Ester w/pad Air Sampling MCE Filters with Pads, 37 mm, 0.8 µm; 100/Pk
Call For Price
COMPARE
25 Laminated-PTFE Air Sampling Laminated-PTFE Filters, 25 mm, 0.2 µm; 100/Pk
₹7,200.00INR / Pkg of 100
COMPARE
25 Laminated-PTFE Air Sampling Laminated-PTFE Filters, 25 mm, 0.45 µm; 100/Pk
₹27,400.00INR / Pkg of 100
COMPARE
37 PTFE Air Sampling PTFE Filters, 37 mm, 1.0 µm; 100/Pk
₹62,300.00INR / Pkg of 100
COMPARE
25 Hydrophobic-PTFE Air Sampling Hydrophobic-PTFE Filters, 25 mm, 1.0 µm; 100/Pk
₹55,000.00INR / Pkg of 100
Showing 1- 15 of 15