Digi-Sense Thermocouple Wire - 24 Gauge
Make your own extension cables
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
10,400.00 - ₹257,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Length (ft) Max Temperature (° F)
Availability Pricing
COMPARE
J 100 221
₹15,100.00INR / Each
COMPARE
J 100 400
₹15,100.00INR / Each
COMPARE
J 100 900
₹16,300.00INR / Each
COMPARE
K 100 221
₹13,500.00INR / Each
COMPARE
K 100 400
₹19,400.00INR / Each
COMPARE
K 100 900
₹16,300.00INR / Each
COMPARE
T 100 221
₹10,400.00INR / Each
COMPARE
T 100 400
₹19,400.00INR / Each
COMPARE
T 100 900
₹13,500.00INR / Each
COMPARE
K 100 900
₹35,900.00INR / Each
COMPARE
J 1000 221
₹93,100.00INR / Each
COMPARE
J 1000 900
₹175,000.00INR / Each
COMPARE
K 1000 221
₹90,400.00INR / Each
COMPARE
K 1000 400
₹257,000.00INR / Each
COMPARE
K 1000 900
₹171,000.00INR / Each
COMPARE
T 1000 221
₹67,000.00INR / Each
COMPARE
T 1000 400
₹131,000.00INR / Each
COMPARE
T 1000 900
₹168,000.00INR / Each
COMPARE
T 1000 2600
₹121,000.00INR / Each
Showing 1- 19 of 19