Digi-Sense T24-1-507 Thermocouple, Wire, Type-T, 24-Gauge, FEP Insulation, 1000-ft Spool
Representative image only.
Digi-Sense T24-1-507 Thermocouple, Wire, Type-T, 24-Gauge, FEP Insulation, 1000-ft Spool
Digi-Sense | Item: 0854213
Make your own extension cables
₹131,000.00INR / Each
₹131,000.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeT
  • MaterialFluorinated ethylene propylene (FEP)
  • Wire Gauge (Ga)24
  • Error Limit±1.5% reading from -330 to -85°F (-200 to -65°C); ±1.8°F (±1°C) from -85 to 270°F (-65 to 130°C); ±0.75% reading from 270 to 660°F (130 to 350°C)
  • Inner InsulationFluorinated ethylene propylene (FEP)
  • Outer InsulationFluorinated ethylene propylene (FEP)
  • Stranded / SolidSolid
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)1000
  • Size (in)0.056 in x 0.092 in
  • DescriptionThermocouple, Wire, Type-T, 24-Gauge, FEP Insulation, 1000-ft Spool