Digi-Sense Thermocouple Probe Wire, Type T, 24-Gauge, Polyvinyl Insulation, RiPCord, 1000ft
Representative image only.
Digi-Sense Thermocouple Probe Wire, Type T, 24-Gauge, Polyvinyl Insulation, RiPCord, 1000ft
Digi-Sense | Item: 0854238
Make your own extension cables
₹121,000.00INR / Each
₹121,000.00INR / Each
Product Description
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
Specifications
  • Connector TypeT
  • MaterialPolyvinyl chloride (PVC)
  • Wire Gauge (Ga)24
  • Inner InsulationPolyvinyl chloride (PVC)
  • Outer InsulationPolyvinyl chloride (PVC)
  • Stranded / SolidSolid
  • Min Temperature (° F)0
  • Max Temperature (° F)2600
  • Min Temperature (° C)-18
  • Max Temperature (° C)1427
  • Length (ft)1000
  • DescriptionThermocouple Probe Wire, Type T, 24-Gauge, Polyvinyl Insulation, RiPCord, 1000ft