Digi-Sense J24-5-502 Thermocouple, Wire, Type-J, 24-Gauge, Polyvinyl Insulation, 1000-ft Spool
Representative image only.
Digi-Sense J24-5-502 Thermocouple, Wire, Type-J, 24-Gauge, Polyvinyl Insulation, 1000-ft Spool
Digi-Sense | Item: 0854206
Make your own extension cables
₹93,100.00INR / Each
₹93,100.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeJ
  • MaterialPolyvinyl chloride (PVC)
  • Wire Gauge (Ga)24
  • Error Limit±4°F (±2.2°C) from 32 to 545°F (0 to 285°C); ±0.75% reading from 545 to 1400°F (285 to 750°C)
  • Inner InsulationPolyvinyl chloride (PVC)
  • Outer InsulationPolyvinyl chloride (PVC)
  • Stranded / SolidSolid
  • Min Temperature (° F)-20
  • Max Temperature (° F)221
  • Min Temperature (° C)-28
  • Max Temperature (° C)105
  • Length (ft)1000
  • Size (in)0.080 in x 0.130 in
  • DescriptionThermocouple, Wire, Type-J, 24-Gauge, Polyvinyl Insulation, 1000-ft Spool