Digi-Sense Thermocouple Wire Type K 24-Gauge, with Ss Overbraid, 100-ft Spool
Representative image only.
Digi-Sense Thermocouple Wire Type K 24-Gauge, with Ss Overbraid, 100-ft Spool
Digi-Sense | Item: 0854136
Make your own extension cables
₹35,900.00INR / Each
₹35,900.00INR / Each
Product Description
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
Specifications
  • Connector TypeK
  • MaterialFiberglass
  • Wire Gauge (Ga)24
  • Inner InsulationFiberglass Wrap
  • Outer InsulationFiberglass Wrap
  • Stranded / SolidSolid
  • OverbraidYes
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)100
  • DescriptionThermocouple Wire Type K 24-Gauge, with Ss Overbraid, 100-ft Spool