Digi-Sense Thermocouple Wire - 20 Gauge
Make your own extension cables
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
12,200.00 - ₹1,171,000.00INR / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Length (ft) Material
Availability Pricing
COMPARE
J 100 Polyvinyl chloride (PVC)
₹13,300.00INR / Each
COMPARE
J 100 Fluorinated ethylene propylene (FEP)
₹21,600.00INR / Each
COMPARE
J 100 Fiberglass
₹16,000.00INR / Each
COMPARE
K 100 Polyvinyl chloride (PVC)
₹19,300.00INR / Each
COMPARE
K 100 Fluorinated ethylene propylene (FEP)
₹33,100.00INR / Each
COMPARE
K 100 Fiberglass
₹26,300.00INR / Each
COMPARE
K 100 High Temp Fiberglass
₹28,800.00INR / Each
COMPARE
K 100 Ceramic Fiber
₹91,100.00INR / Each
COMPARE
T 100 Polyvinyl chloride (PVC)
₹12,200.00INR / Each
COMPARE
T 100 Fluorinated ethylene propylene (FEP)
₹24,900.00INR / Each
COMPARE
T 100 Fiberglass
₹26,000.00INR / Each
COMPARE
J 100 Fiberglass
₹38,200.00INR / Each
COMPARE
K 100 Fiberglass
₹44,000.00INR / Each
COMPARE
J 100 Polyvinyl chloride (PVC)
₹13,300.00INR / Each
COMPARE
J 1000 Polyvinyl chloride (PVC)
₹125,000.00INR / Each
COMPARE
J 1000 Fluorinated ethylene propylene (FEP)
₹258,000.00INR / Each
COMPARE
J 1000 Fiberglass
₹150,000.00INR / Each
COMPARE
K 1000 Polyvinyl chloride (PVC)
₹223,000.00INR / Each
COMPARE
K 1000 Fluorinated ethylene propylene (FEP)
₹348,000.00INR / Each
COMPARE
K 1000 Fiberglass
₹285,000.00INR / Each
Showing 1 - 20 of 24