Digi-Sense J20-5-502 Thermocouple, Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 100-ft Spool
Representative image only.
Digi-Sense J20-5-502 Thermocouple, Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 100-ft Spool
Digi-Sense | Item: 0854116
Make your own extension cables
₹13,300.00INR / Each
₹13,300.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeJ
  • MaterialPolyvinyl chloride (PVC)
  • Wire Gauge (Ga)20
  • Error Limit±4°F (±2.2°C) from 32 to 545°F (0 to 285°C); ±0.75% reading from 545 to 1400°F (285 to 750°C)
  • Inner InsulationPolyvinyl chloride (PVC)
  • Outer InsulationPolyvinyl chloride (PVC)
  • Stranded / SolidSolid
  • OverbraidNo
  • Min Temperature (° F)-20
  • Max Temperature (° F)221
  • Min Temperature (° C)-28
  • Max Temperature (° C)105
  • Length (ft)100
  • Size (in)0.092 in x 0.154 in
  • DescriptionThermocouple, Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 100-ft Spool